Product Details
- SNP ID
-
rs143501297
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:178149741 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGGGTAGGGGCAGGGACTGCACC[G/T]CCTCCAGGCACTCATCGTAAGCCTC
- Phenotype
-
MIM: 606470
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NHP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147575606] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NHP2
- Gene Name
- NHP2 ribonucleoprotein
- Gene
- RMND5B
- Gene Name
- required for meiotic nuclear division 5 homolog B
There are no transcripts associated with this gene.
View Full Product Details