Product Details

SNP ID
rs143543627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160349661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTTGGGCCCCTTCTTGCCTGGT[A/G]TGCCATGCTTCCCGGGGGTACCGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF2 PubMed Links

Gene Details

Gene
C1QTNF2
Gene Name
C1q and tumor necrosis factor related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031908.4 636 Missense Mutation ACA,ATA T167I NP_114114.2
XM_011534425.2 636 Missense Mutation ACA,ATA T206I XP_011532727.1
XM_011534426.2 636 Intron XP_011532728.1
XM_011534427.2 636 Intron XP_011532729.1
XM_017009013.1 636 Intron XP_016864502.1

View Full Product Details