Product Details

SNP ID
rs143688606
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:58456088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTATCATCCGAATAGCATCCCCA[A/G]TCTGCTGCTTGTTTTCTACTGCGGG
Phenotype
MIM: 607023
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PLK2 PubMed Links

Gene Details

Gene
PLK2
Gene Name
polo like kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252226.1 1502 Missense Mutation ACT,ATT T427I NP_001239155.1
NM_006622.3 1502 Missense Mutation ACT,ATT T441I NP_006613.2

View Full Product Details