Product Details

SNP ID
rs143939877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150168902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAAAGGTACAAATGTTGCATCA[C/T]TGGGCAATTTCAGGGGTTCTGGAGG
Phenotype
MIM: 600746
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDX1 PubMed Links

Gene Details

Gene
CDX1
Gene Name
caudal type homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001804.2 Intron NP_001795.2

View Full Product Details