Product Details

SNP ID
rs144080763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:39106822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCATACTAAATGCATAACATTC[A/G]TATCAAATAATTAACATTTATATAC
Phenotype
MIM: 602731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FYB PubMed Links

Gene Details

Gene
FYB
Gene Name
FYN binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243093.1 3097 UTR 3 NP_001230022.1
NM_001465.4 3097 UTR 3 NP_001456.3
NM_199335.3 3097 UTR 3 NP_955367.1
XM_006714464.2 3097 UTR 3 XP_006714527.1
XM_006714465.2 3097 UTR 3 XP_006714528.1
XM_006714466.2 3097 UTR 3 XP_006714529.1
XM_011514008.2 3097 UTR 3 XP_011512310.1
XM_011514009.1 3097 UTR 3 XP_011512311.1
XM_011514010.1 3097 UTR 3 XP_011512312.1
XM_011514011.2 3097 UTR 3 XP_011512313.1
XM_011514012.2 3097 Intron XP_011512314.1
XM_011514013.2 3097 Intron XP_011512315.1
XM_017009316.1 3097 UTR 3 XP_016864805.1

View Full Product Details