Product Details

SNP ID
rs144304574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:160252524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTCCAAGACGTGGAATCTTCC[A/G]GAACCGAGAGCTGGGAAAATGGTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNJL PubMed Links

Gene Details

Gene
CCNJL
Gene Name
cyclin J like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308173.1 2083 UTR 3 NP_001295102.1
NM_024565.6 2083 UTR 3 NP_078841.3
XM_006714917.3 2083 UTR 3 XP_006714980.1
XM_011534646.2 2083 UTR 3 XP_011532948.1
XM_011534647.2 2083 UTR 3 XP_011532949.1
XM_017009846.1 2083 UTR 3 XP_016865335.1
XM_017009847.1 2083 UTR 3 XP_016865336.1
XM_017009848.1 2083 UTR 3 XP_016865337.1
XM_017009849.1 2083 UTR 3 XP_016865338.1

View Full Product Details