Product Details

SNP ID
rs144324241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:136129513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAATAACACAGAATTCTCAGAA[C/T]TTGGAAAGGAAGCTTTATTGTTTCC
Phenotype
MIM: 603110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMAD5 PubMed Links

Gene Details

Gene
SMAD5
Gene Name
SMAD family member 5
There are no transcripts associated with this gene.

Gene
SMAD5-AS1
Gene Name
SMAD5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details