Product Details
- SNP ID
-
rs144759983
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:40841477 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGATCCTGATTCTATCTTAGACA[C/T]GTTAACTTCTCGGAGGCTGATTTCT
- Phenotype
-
MIM: 609986
MIM: 604181
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CARD6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141244584] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CARD6
- Gene Name
- caspase recruitment domain family member 6
- Gene
- RPL37
- Gene Name
- ribosomal protein L37
There are no transcripts associated with this gene.
- Gene
- SNORD72
- Gene Name
- small nucleolar RNA, C/D box 72
There are no transcripts associated with this gene.
View Full Product Details