Product Details

SNP ID
rs144777583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149995269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGTGGATGGCTTGACTGACCA[C/T]CACGCTGCAGCAATCATGTCGATGG
Phenotype
MIM: 606718
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMGXB3 PubMed Links

Gene Details

Gene
HMGXB3
Gene Name
HMG-box containing 3
There are no transcripts associated with this gene.

Gene
SLC26A2
Gene Name
solute carrier family 26 member 2
There are no transcripts associated with this gene.

Gene
TIGD6
Gene Name
tigger transposable element derived 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243253.1 1855 Nonsense Mutation TGA,TGG *360W NP_001230182.1
NM_030953.3 1855 Nonsense Mutation TGA,TGG *360W NP_112215.1

View Full Product Details