Product Details

SNP ID
rs144815922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140566807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGATGAGCAGCTCAGCCTTCA[A/G]GTGGAGACACTTTATCTGGATTCCC
Phenotype
MIM: 602711 MIM: 603819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APBB3 PubMed Links

Gene Details

Gene
APBB3
Gene Name
amyloid beta precursor protein binding family B member 3
There are no transcripts associated with this gene.

Gene
MIR6831
Gene Name
microRNA 6831
There are no transcripts associated with this gene.

Gene
SLC35A4
Gene Name
solute carrier family 35 member A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080670.3 636 UTR 5 NP_542401.1
Gene
SRA1
Gene Name
steroid receptor RNA activator 1
There are no transcripts associated with this gene.

View Full Product Details