Product Details

SNP ID
rs144865261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141101032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTCCGTATCATAGATGTAAATG[A/G]CCATTCTCCGGTATTCTTTGAAAAT
Phenotype
MIM: 606328 MIM: 606329
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB2 PubMed Links

Gene Details

Gene
PCDHB2
Gene Name
protocadherin beta 2
There are no transcripts associated with this gene.

Gene
PCDHB3
Gene Name
protocadherin beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018937.4 791 Missense Mutation GAC,GGC D128G NP_061760.1

View Full Product Details