Product Details
- SNP ID
-
hCV171385004
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:140807597 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCCGCCGGTGTTCCCAGCAACAC[A/C]AAAGAACCTGTCCATCGCGGAATCC
- Phenotype
-
MIM: 606307
MIM: 606308
MIM: 606309
MIM: 606310
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PCDHA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3822346] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PCDHA1
- Gene Name
- protocadherin alpha 1
There are no transcripts associated with this gene.
- Gene
- PCDHA2
- Gene Name
- protocadherin alpha 2
There are no transcripts associated with this gene.
- Gene
- PCDHA3
- Gene Name
- protocadherin alpha 3
There are no transcripts associated with this gene.
- Gene
- PCDHA4
- Gene Name
- protocadherin alpha 4
There are no transcripts associated with this gene.
View Full Product Details