Product Details

SNP ID
rs145263618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:67183178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAGACGGGGTGAGATGATGTT[A/G]ATGCTGTTGGCAGCCAGATTGAGGT
Phenotype
MIM: 602226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD180 PubMed Links

Gene Details

Gene
CD180
Gene Name
CD180 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005582.2 1822 Silent Mutation ATC,ATT I555I NP_005573.2
XM_005248504.4 1822 Silent Mutation ATC,ATT I542I XP_005248561.1

View Full Product Details