Product Details

SNP ID
rs145474099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138512809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTAGTTCAGTTTTAAAGTCAGC[A/G]GATCCAGCTAAAACTAGACCAGCCA
Phenotype
MIM: 600285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETF1 PubMed Links

Gene Details

Gene
ETF1
Gene Name
eukaryotic translation termination factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256302.1 1108 Silent Mutation TCC,TCT S196S NP_001243231.1
NM_001282185.1 1108 Silent Mutation TCC,TCT S215S NP_001269114.1
NM_001291974.1 1108 Silent Mutation TCC,TCT S196S NP_001278903.1
NM_001291975.1 1108 Silent Mutation TCC,TCT S196S NP_001278904.1
NM_004730.3 1108 Silent Mutation TCC,TCT S229S NP_004721.1
XM_005271921.1 1108 Silent Mutation TCC,TCT S196S XP_005271978.1
XM_017009221.1 1108 Silent Mutation TCC,TCT S229S XP_016864710.1
XM_017009222.1 1108 Intron XP_016864711.1

View Full Product Details