Product Details

SNP ID
rs145551245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:40843196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGTACCTCCTCAATCTATGGGG[A/G]CAAGCAGTAATTCAGAAGATGCTTT
Phenotype
MIM: 609986 MIM: 604181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CARD6 PubMed Links

Gene Details

Gene
CARD6
Gene Name
caspase recruitment domain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032587.3 403 Missense Mutation ACA,GCA T110A NP_115976.2
XM_017009989.1 403 Intron XP_016865478.1
XM_017009990.1 403 UTR 5 XP_016865479.1
Gene
RPL37
Gene Name
ribosomal protein L37
There are no transcripts associated with this gene.

View Full Product Details