Product Details

SNP ID
rs145584571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157162804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCCATGTCGCTTGCCCCTTAC[A/G]TTGCTGTACCCTTTTTCTTTTTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM71B PubMed Links

Gene Details

Gene
FAM71B
Gene Name
family with sequence similarity 71 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130899.2 1561 Silent Mutation AAC,AAT N487N NP_570969.2

View Full Product Details