Product Details

SNP ID
rs145828177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:60599178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGAGAGCCATATGATCTTTGA[C/T]ATTCAAATTCCTCTGGACTAATTTG
Phenotype
MIM: 616073
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEPDC1B PubMed Links

Gene Details

Gene
DEPDC1B
Gene Name
DEP domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145208.1 1391 Intron NP_001138680.1
NM_018369.2 1391 Missense Mutation TAT,TGT Y442C NP_060839.2
XM_011543509.2 1391 Missense Mutation TAT,TGT Y427C XP_011541811.1
XM_011543510.2 1391 Intron XP_011541812.1
XM_017009629.1 1391 Intron XP_016865118.1

View Full Product Details