Product Details

SNP ID
rs145858925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:147852541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAAGCCTAGGAAGAAAGTCAC[C/T]GTGTAGATGATCATGGAAAAGATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf46 PubMed Links

Gene Details

Gene
C5orf46
Gene Name
chromosome 5 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206966.2 Intron NP_996849.2
XM_005268446.3 Intron XP_005268503.2
XM_017009459.1 Intron XP_016864948.1

View Full Product Details