Product Details

SNP ID
rs145898895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:67183428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTCCCTTTTAAGTTGAGATGC[C/T]GGAGAACTGGTAGGCCTGCTAGAAG
Phenotype
MIM: 602226
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD180 PubMed Links

Gene Details

Gene
CD180
Gene Name
CD180 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005582.2 1572 Missense Mutation CAG,CGG Q472R NP_005573.2
XM_005248504.4 1572 Missense Mutation CAG,CGG Q459R XP_005248561.1

View Full Product Details