Product Details

SNP ID
rs145994010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:137753848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCTTCAGCTCCACAGTGTTGCC[A/G]TCCACGGCATGGGGCGAGGCGGCCA
Phenotype
MIM: 609409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNRNPA0 PubMed Links

Gene Details

Gene
HNRNPA0
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006805.3 503 Silent Mutation GAC,GAT D73D NP_006796.1

View Full Product Details