Product Details

SNP ID
rs146055360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:54455832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCACCTCATAGAGATTCCAGTG[C/T]GTTACCAGGAAGAGTTTGAAGCTCG
Phenotype
MIM: 604624
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSPB3 PubMed Links

Gene Details

Gene
HSPB3
Gene Name
heat shock protein family B (small) member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006308.2 232 Missense Mutation CGT,TGT R15C NP_006299.1

View Full Product Details