Product Details

SNP ID
rs146107880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177457746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCTGTGATTGACTGAAGTA[C/G]CCCTCACTGGCCTGCAGGGGAAAGC
Phenotype
MIM: 126660
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DBN1 PubMed Links

Gene Details

Gene
DBN1
Gene Name
drebrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004395.3 2076 Silent Mutation GGC,GGG G596G NP_004386.2
NM_080881.2 2076 Silent Mutation GGC,GGG G598G NP_543157.1
XM_005265827.3 2076 Silent Mutation GGC,GGG G642G XP_005265884.1
XM_006714826.2 2076 Silent Mutation GGC,GGG G579G XP_006714889.1
XM_011534446.2 2076 Silent Mutation GGC,GGG G644G XP_011532748.1
XM_011534447.2 2076 Silent Mutation GGC,GGG G631G XP_011532749.1
XM_017009139.1 2076 Silent Mutation GGC,GGG G644G XP_016864628.1
XM_017009140.1 2076 Silent Mutation GGC,GGG G598G XP_016864629.1
Gene
PRR7
Gene Name
proline rich 7 (synaptic)
There are no transcripts associated with this gene.

Gene
PRR7-AS1
Gene Name
PRR7 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details