Product Details

SNP ID
rs146187959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141330771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCTGTTGGAAGCTGGGGCTGG[A/G]AATATTCACTACTCAGTGCCGGAAG
Phenotype
MIM: 606288 MIM: 606289 MIM: 600573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 87 Silent Mutation GGA,GGG G29G NP_061735.1
NM_031993.1 87 Silent Mutation GGA,GGG G29G NP_114382.1
Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
There are no transcripts associated with this gene.

Gene
TAF7
Gene Name
TATA-box binding protein associated factor 7
There are no transcripts associated with this gene.

View Full Product Details