Product Details

SNP ID
rs146382764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1201806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCTGGGCTTCTGCGTGGGCCT[C/T]GGCAACGTGTGGCGCTTCCCCTACC
Phenotype
MIM: 608893
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A19 PubMed Links

Gene Details

Gene
SLC6A19
Gene Name
solute carrier family 6 member 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003841.2 212 Silent Mutation CTC,CTT L52L NP_001003841.1

View Full Product Details