Product Details

SNP ID
rs146407384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:65670264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTAAATCAGGAATGCTCTTCAGC[A/G]CTGCTGCTGAATGATCTGCTCCGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGTB PubMed Links

Gene Details

Gene
SGTB
Gene Name
small glutamine rich tetratricopeptide repeat containing beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019072.2 1253 Silent Mutation AGC,AGT S299S NP_061945.1
XM_005248548.3 1253 Silent Mutation AGC,AGT S299S XP_005248605.1
XM_017009597.1 1253 Silent Mutation AGC,AGT S234S XP_016865086.1
Gene
TRAPPC13
Gene Name
trafficking protein particle complex 13
There are no transcripts associated with this gene.

View Full Product Details