Product Details
- SNP ID
-
rs146479455
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:39107463 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTGCTAGTCATTGTCATAGATGC[A/G]GCCTGAAAGGAAAAAATACAAATTT
- Phenotype
-
MIM: 602731
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FYB
PubMed Links
Gene Details
- Gene
- FYB
- Gene Name
- FYN binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001243093.1 |
2456 |
Missense Mutation |
CGC,TGC |
R834C |
NP_001230022.1 |
NM_001465.4 |
2456 |
Missense Mutation |
CGC,TGC |
R824C |
NP_001456.3 |
NM_199335.3 |
2456 |
Missense Mutation |
CGC,TGC |
R778C |
NP_955367.1 |
XM_006714464.2 |
2456 |
Missense Mutation |
CGC,TGC |
R824C |
XP_006714527.1 |
XM_006714465.2 |
2456 |
Missense Mutation |
CGC,TGC |
R824C |
XP_006714528.1 |
XM_006714466.2 |
2456 |
Missense Mutation |
CGC,TGC |
R824C |
XP_006714529.1 |
XM_011514008.2 |
2456 |
Missense Mutation |
CGC,TGC |
R834C |
XP_011512310.1 |
XM_011514009.1 |
2456 |
Missense Mutation |
CGC,TGC |
R824C |
XP_011512311.1 |
XM_011514010.1 |
2456 |
Missense Mutation |
CGC,TGC |
R824C |
XP_011512312.1 |
XM_011514011.2 |
2456 |
Missense Mutation |
CGC,TGC |
R788C |
XP_011512313.1 |
XM_011514012.2 |
2456 |
Intron |
|
|
XP_011512314.1 |
XM_011514013.2 |
2456 |
Intron |
|
|
XP_011512315.1 |
XM_017009316.1 |
2456 |
Missense Mutation |
CGC,TGC |
R788C |
XP_016864805.1 |
View Full Product Details