Product Details

SNP ID
rs146674681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1879579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCTCCTCCTCGCCCTCCGCGT[A/G]GGGCCGCTTCTCGTCTGCGCACTTG
Phenotype
MIM: 606199
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTD-2194D22.4 PubMed Links

Gene Details

Gene
CTD-2194D22.4
Gene Name
uncharacterized LOC101929081
There are no transcripts associated with this gene.

Gene
IRX4
Gene Name
iroquois homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278632.1 879 Missense Mutation NP_001265561.1
NM_001278633.1 879 Missense Mutation NP_001265562.1
NM_001278634.1 879 Missense Mutation NP_001265563.1
NM_001278635.1 879 Missense Mutation NP_001265564.1
NM_016358.2 879 Missense Mutation NP_057442.1

View Full Product Details