Product Details

SNP ID
rs146889379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79980759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCTCACAGTATTCTGTTTAAGA[C/T]TGTGGACCATTTATCCCAATGAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTX3 PubMed Links

Gene Details

Gene
MTX3
Gene Name
metaxin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010891.4 3726 UTR 3 NP_001010891.4
NM_001167741.1 3726 UTR 3 NP_001161213.1
XM_005248495.3 3726 UTR 3 XP_005248552.1
XM_017009440.1 3726 UTR 3 XP_016864929.1

View Full Product Details