Product Details
- SNP ID
-
rs147041695
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:170878122 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTTGGCTGAATTGGAAGTGTTAT[A/G]TACTCATCTCTACATAGGGACTGAT
- Phenotype
-
MIM: 606141
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RANBP17
PubMed Links
Gene Details
- Gene
- RANBP17
- Gene Name
- RAN binding protein 17
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022897.4 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
NP_075048.1 |
XM_011534627.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_011532929.1 |
XM_011534631.2 |
52 |
Missense Mutation |
TAT,TGT |
Y17C |
XP_011532933.1 |
XM_011534636.2 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_011532938.1 |
XM_011534637.2 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_011532939.1 |
XM_017009736.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_016865225.1 |
XM_017009737.1 |
52 |
Missense Mutation |
TAT,TGT |
Y17C |
XP_016865226.1 |
XM_017009738.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_016865227.1 |
XM_017009739.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_016865228.1 |
XM_017009740.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_016865229.1 |
XM_017009741.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_016865230.1 |
XM_017009742.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_016865231.1 |
XM_017009743.1 |
52 |
UTR 5 |
|
|
XP_016865232.1 |
XM_017009744.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_016865233.1 |
XM_017009745.1 |
52 |
Intron |
|
|
XP_016865234.1 |
XM_017009746.1 |
52 |
Missense Mutation |
TAT,TGT |
Y15C |
XP_016865235.1 |
XM_017009747.1 |
52 |
Intron |
|
|
XP_016865236.1 |
View Full Product Details