Product Details

SNP ID
rs147674827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141331211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCTACCAACTCAGCTCTAACC[C/T]TCATTTCTCCCTGGATGTGCAACAG
Phenotype
MIM: 606288 MIM: 606289
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 527 Missense Mutation CCT,CTT P176L NP_061735.1
NM_031993.1 527 Missense Mutation CCT,CTT P176L NP_114382.1
Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
There are no transcripts associated with this gene.

View Full Product Details