Product Details

SNP ID
rs147804421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95566085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGACACGTGGGACTTCCTTTCTG[A/T]ATGCCTACACAAACTCTCCAATTTG
Phenotype
MIM: 610011
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARSK PubMed Links

Gene Details

Gene
ARSK
Gene Name
arylsulfatase family member K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198150.2 372 Missense Mutation AAT,TAT N72Y NP_937793.1
XM_005271904.3 372 UTR 5 XP_005271961.1

View Full Product Details