Product Details

SNP ID
rs148015838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:54518702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCGTGCTGGGGGAGGCGTCAGG[C/T]TTCGTGAAGGACGGGGACAAGCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNX18 PubMed Links

Gene Details

Gene
SNX18
Gene Name
sorting nexin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102575.1 940 Silent Mutation GGC,GGT G250G NP_001096045.1
NM_001145427.1 940 Silent Mutation GGC,GGT G250G NP_001138899.1
NM_052870.2 940 Silent Mutation GGC,GGT G250G NP_443102.2
XM_017008997.1 940 Silent Mutation GGC,GGT G250G XP_016864486.1

View Full Product Details