Product Details

SNP ID
rs148257148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140335934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGACAGCACCACAGCCACCAC[A/G]GCCAGGATGGTTGTGTGGTCATAGG
Phenotype
MIM: 126150
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HBEGF PubMed Links

Gene Details

Gene
HBEGF
Gene Name
heparin binding EGF like growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001945.2 767 Silent Mutation GCC,GCT A164A NP_001936.1

View Full Product Details