Product Details

SNP ID
rs148324701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157138824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATGACCTGAATTTTCTCTTTGA[C/T]GTTCATTCTGTCCAGTACAATTGTT
Phenotype
MIM: 605045
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED7 PubMed Links

Gene Details

Gene
MED7
Gene Name
mediator complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100816.1 716 Missense Mutation NP_001094286.1
NM_004270.4 716 Missense Mutation NP_004261.1

View Full Product Details