Product Details

SNP ID
rs148536759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139661979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGGGGAGAGATACAGGGGAGCT[A/G]GGAAAAGGAAAGGCTGGAGAAGGAG
Phenotype
MIM: 612752
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXXC5 PubMed Links

Gene Details

Gene
CXXC5
Gene Name
CXXC finger protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317199.1 Intron NP_001304128.1
NM_001317200.1 Intron NP_001304129.1
NM_001317201.1 Intron NP_001304130.1
NM_001317202.1 Intron NP_001304131.1
NM_001317203.1 Intron NP_001304132.1
NM_001317204.1 Intron NP_001304133.1
NM_001317205.1 Intron NP_001304134.1
NM_001317206.1 Intron NP_001304135.1
NM_001317207.1 Intron NP_001304136.1
NM_001317208.1 Intron NP_001304137.1
NM_001317209.1 Intron NP_001304138.1
NM_001317210.1 Intron NP_001304139.1
NM_001317211.1 Intron NP_001304140.1
NM_016463.8 Intron NP_057547.5
XM_017009573.1 Intron XP_016865062.1

View Full Product Details