Product Details

SNP ID
rs148548964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69192095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGACACAGGGTTTTGCCACCACA[C/T]CTGGCCTCAAATAAGATTTTGTGAG
Phenotype
MIM: 605607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPH PubMed Links

Gene Details

Gene
CENPH
Gene Name
centromere protein H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022909.3 Intron NP_075060.1

View Full Product Details