Product Details

SNP ID
rs148551532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64507851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCTGTGAAATGGCCCGTCAGGC[A/G]CACCAAAAATTGGCTGCCATCTCAG
Phenotype
MIM: 610890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS7BP PubMed Links

Gene Details

Gene
RGS7BP
Gene Name
regulator of G-protein signaling 7 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029875.2 2366 Silent Mutation GCA,GCG A102A NP_001025046.1
NM_001271890.1 2366 Intron NP_001258819.1
NM_001271891.1 2366 Intron NP_001258820.1
XM_005248502.3 2366 Silent Mutation GCA,GCG A102A XP_005248559.1

View Full Product Details