Product Details
- SNP ID
-
rs148668212
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:141180293 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTGTCTCAGAACTATCAGTACGA[A/G]GTGTGCCTGGCAGGAGGCTCAGGGA
- Phenotype
-
MIM: 606345
MIM: 606333
MIM: 606334
MIM: 606335
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PCDHB16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17096961] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PCDHB16
- Gene Name
- protocadherin beta 16
There are no transcripts associated with this gene.
- Gene
- PCDHB7
- Gene Name
- protocadherin beta 7
There are no transcripts associated with this gene.
- Gene
- PCDHB8
- Gene Name
- protocadherin beta 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019120.4 |
2504 |
Silent Mutation |
GAA,GAG |
E753E |
NP_061993.3 |
- Gene
- PCDHB9
- Gene Name
- protocadherin beta 9
There are no transcripts associated with this gene.
View Full Product Details