Product Details

SNP ID
rs148748297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:181255983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCATTTAGATCTTCAAAAAGCAG[A/G]GCAAACTGGAATGGAATGTACTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTC-338M12.4 PubMed Links

Gene Details

Gene
CTC-338M12.4
Gene Name
uncharacterized LOC101928649
There are no transcripts associated with this gene.

Gene
TRIM52
Gene Name
tripartite motif containing 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032765.2 2406 Intron NP_116154.1
XM_005266000.3 2406 UTR 3 XP_005266057.1
XM_017009991.1 2406 Intron XP_016865480.1
XM_017009992.1 2406 UTR 3 XP_016865481.1
XM_017009993.1 2406 UTR 3 XP_016865482.1
XM_017009994.1 2406 UTR 3 XP_016865483.1
Gene
TRIM52-AS1
Gene Name
TRIM52 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details