Product Details

SNP ID
rs148830499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:146339659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAACCATCCGTTCAAGCCCGAC[A/G]CCACCTACCATACCATGAGCAGCGT
Phenotype
MIM: 602460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POU4F3 PubMed Links

Gene Details

Gene
POU4F3
Gene Name
POU class 4 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002700.2 321 Missense Mutation ACC,GCC T78A NP_002691.1

View Full Product Details