Product Details

SNP ID
rs148849408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:32126449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCATTCACCCATTTGTCAAGAAC[A/G]GCTTCCTGTACTTTCTTGATGAGGC
Phenotype
MIM: 612207
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLPH3 PubMed Links

Gene Details

Gene
GOLPH3
Gene Name
golgi phosphoprotein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022130.3 1214 Silent Mutation GCC,GCT A220A NP_071413.1
XM_017009686.1 1214 Silent Mutation GCC,GCT A128A XP_016865175.1
XM_017009687.1 1214 Silent Mutation GCC,GCT A128A XP_016865176.1

View Full Product Details