Product Details

SNP ID
rs148900904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131651947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACTTGAACAGTCCATTTATCC[A/G]TGTCAGCTATAATACAGACAGCTTC
Phenotype
MIM: 610594
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNIP1 PubMed Links

Gene Details

Gene
FNIP1
Gene Name
folliculin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008738.2 3219 Missense Mutation ACG,ATG T1026M NP_001008738.2
NM_133372.2 3219 Missense Mutation ACG,ATG T1054M NP_588613.2

View Full Product Details