Product Details
- SNP ID
-
rs149026271
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:151183553 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCCTTCTCCTGCTGGAGCTGTC[A/G]CCGCAGCTGCACCTCCTTCTCCAGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC69
PubMed Links
Gene Details
- Gene
- CCDC69
- Gene Name
- coiled-coil domain containing 69
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015621.2 |
898 |
Nonsense Mutation |
CGA,TGA |
R259* |
NP_056436.2 |
View Full Product Details