Product Details
- SNP ID
-
rs149148720
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:138140730 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAATCTGCTCCAGGACTTCTTGC[C/T]GCATCTCCACAGCCATATGGTATAC
- Phenotype
-
MIM: 602848
MIM: 603575
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BRD8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs412051] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BRD8
- Gene Name
- bromodomain containing 8
- Gene
- NME5
- Gene Name
- NME/NM23 family member 5
There are no transcripts associated with this gene.
View Full Product Details