Product Details

SNP ID
rs149148884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176392927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCTCCTCCTTGGATTCCTTCAC[A/G]AAAGCCTCCTCGGATGCCCTGCGGG
Phenotype
MIM: 118970 MIM: 612861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL10 PubMed Links

Gene Details

Gene
ARL10
Gene Name
ADP ribosylation factor like GTPase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317948.1 688 Intron NP_001304877.1
NM_173664.5 688 Intron NP_775935.1
XM_011534529.2 688 Intron XP_011532831.1
XM_011534530.2 688 Intron XP_011532832.1
XM_011534531.2 688 Intron XP_011532833.1
XM_017009372.1 688 Intron XP_016864861.1
Gene
CLTB
Gene Name
clathrin light chain B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001834.3 688 Silent Mutation TTC,TTT F161F NP_001825.1
NM_007097.3 688 Silent Mutation TTC,TTT F179F NP_009028.1
XM_017009026.1 688 Silent Mutation TTC,TTT F94F XP_016864515.1
XM_017009027.1 688 Intron XP_016864516.1
Gene
HIGD2A
Gene Name
HIG1 hypoxia inducible domain family member 2A
There are no transcripts associated with this gene.

Gene
NOP16
Gene Name
NOP16 nucleolar protein
There are no transcripts associated with this gene.

View Full Product Details