Product Details

SNP ID
rs149174030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73123575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGTGGCGGGGCCTGCATGTGCC[A/G]TGGTTGGGCTTGGGGCTGTGATCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105379030 PubMed Links

Gene Details

Gene
LOC105379030
Gene Name
uncharacterized LOC105379030
There are no transcripts associated with this gene.

Gene
TMEM171
Gene Name
transmembrane protein 171
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161342.2 406 Missense Mutation ATG,GTG M68V NP_001154814.1
NM_173490.7 406 Missense Mutation ATG,GTG M68V NP_775761.4
XM_011543156.1 406 Missense Mutation ATG,GTG M68V XP_011541458.1

View Full Product Details