Product Details

SNP ID
rs149209328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75625897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCCACTCAGAGCAATCATGTGC[A/G]CATCGTGGAGTATCTTATTCAAGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKDD1B PubMed Links

Gene Details

Gene
ANKDD1B
Gene Name
ankyrin repeat and death domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276713.1 993 Missense Mutation CAC,CGC H181R NP_001263642.1
XM_011543618.2 993 Missense Mutation CAC,CGC H181R XP_011541920.1
XM_011543623.2 993 Intron XP_011541925.1
XM_017009814.1 993 Missense Mutation CAC,CGC H216R XP_016865303.1
XM_017009815.1 993 Missense Mutation CAC,CGC H216R XP_016865304.1
XM_017009816.1 993 Missense Mutation CAC,CGC H216R XP_016865305.1
XM_017009817.1 993 Intron XP_016865306.1

View Full Product Details