Product Details

SNP ID
rs149402782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157162455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCCCTGGCTCCTCTTTAAATGG[A/G]TTTGGTTTCAAAGACGATCATCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM71B PubMed Links

Gene Details

Gene
FAM71B
Gene Name
family with sequence similarity 71 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130899.2 1910 Missense Mutation CCC,TCC P604S NP_570969.2

View Full Product Details