Product Details
- SNP ID
-
rs149470983
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:111501009 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATGTTGAGATTATTACCACAAGA[C/T]AAAAGCCCTTCTTTATAGCCCACAG
- Phenotype
-
MIM: 114080
MIM: 607049
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAMK4
PubMed Links
Gene Details
- Gene
- CAMK4
- Gene Name
- calcium/calmodulin dependent protein kinase IV
There are no transcripts associated with this gene.
- Gene
- STARD4
- Gene Name
- StAR related lipid transfer domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308056.1 |
612 |
Silent Mutation |
TTA,TTG |
L130L |
NP_001294985.1 |
NM_001308057.1 |
612 |
Silent Mutation |
TTA,TTG |
L130L |
NP_001294986.1 |
NM_001308058.1 |
612 |
Silent Mutation |
TTA,TTG |
L130L |
NP_001294987.1 |
NM_001308059.1 |
612 |
Silent Mutation |
TTA,TTG |
L48L |
NP_001294988.1 |
NM_001308060.1 |
612 |
Silent Mutation |
TTA,TTG |
L32L |
NP_001294989.1 |
NM_001308061.1 |
612 |
Silent Mutation |
TTA,TTG |
L48L |
NP_001294990.1 |
NM_139164.2 |
612 |
Silent Mutation |
TTA,TTG |
L130L |
NP_631903.1 |
XM_017009041.1 |
612 |
Silent Mutation |
TTA,TTG |
L32L |
XP_016864530.1 |
XM_017009042.1 |
612 |
Silent Mutation |
TTA,TTG |
L32L |
XP_016864531.1 |
XM_017009043.1 |
612 |
Silent Mutation |
TTA,TTG |
L32L |
XP_016864532.1 |
XM_017009044.1 |
612 |
Silent Mutation |
TTA,TTG |
L48L |
XP_016864533.1 |
View Full Product Details