Product Details

SNP ID
rs149546549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:126601017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGCTTTCCGACTCGGATTCC[G/T]ACATGACGGTCGCACCCAGCGACAG
Phenotype
MIM: 107323 MIM: 604924
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH7A1 PubMed Links

Gene Details

Gene
ALDH7A1
Gene Name
aldehyde dehydrogenase 7 family member A1
There are no transcripts associated with this gene.

Gene
PHAX
Gene Name
phosphorylated adaptor for RNA export
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032177.3 Intron NP_115553.2

View Full Product Details